Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257184.2 | XP_008255406.1 | 6 | 1849 | 108 | |
| XM_008257185.2 | XP_008255407.1 | 7 | 1889 | 108 | ||
| XM_008257186.2 | XP_008255408.1 | 6 | 1122 | 108 | ||
| XM_008257187.2 | XP_008255409.1 | 6 | 1747 | 108 | ||
More Information
Set Membership: |
|

ZNF570
Genomic Map