Assay Details
| Gene Symbol: LOC100359126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258111.2 | XP_008256333.2 | 7 - 8 | 2287 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100359126| Gene Symbol: LOC100359126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258111.2 | XP_008256333.2 | 7 - 8 | 2287 | 113 | |
Set Membership: |
|