Assay Details
| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258399.1 | XP_008256621.1 | 8 - 9 | 914 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX4| Gene Symbol: PAX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258399.1 | XP_008256621.1 | 8 - 9 | 914 | 57 | |
Set Membership: |
|