Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262333.2 | XP_008260555.1 | 1 - 2 | 138 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008262333.2 transcripts.
Set Membership: |
|

CCL28
Genomic Map