Assay Details
| Gene Symbol: LOC103345260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337856.1 | XP_017193345.1 | 1890 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103345260| Gene Symbol: LOC103345260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017337856.1 | XP_017193345.1 | 1890 | 98 | ||
Set Membership: |
|