Assay Details
Gene Symbol: LOC100341315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017338150.1 | XP_017193639.1 | 1524 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
