Assay Details
| Gene Symbol: LOC103345648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338690.1 | XP_017194179.1 | 1344 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103345648| Gene Symbol: LOC103345648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338690.1 | XP_017194179.1 | 1344 | 82 | ||
Set Membership: |
|