Assay Details
| Gene Symbol: LOC108177042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343593.1 | XP_017199082.1 | 3 - 4 | 1452 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108177042| Gene Symbol: LOC108177042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343593.1 | XP_017199082.1 | 3 - 4 | 1452 | 64 | |
Set Membership: |
|