Assay Details
| Gene Symbol: LOC100339790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708421.2 | XP_002708467.1 | 1 - 2 | 136 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100339790| Gene Symbol: LOC100339790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708421.2 | XP_002708467.1 | 1 - 2 | 136 | 100 | |
Set Membership: |
|