Assay Details
| Gene Symbol: SP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711016.3 | XP_002711062.1 | 2 - 3 | 367 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002711016.3 transcripts.
Set Membership: |
|

SP7
Genomic Map