Assay Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712170.3 | XP_002712216.1 | 2 - 3 | 330 | 66 | |
| XM_008258705.2 | XP_008256927.1 | 2 - 3 | 330 | 66 | ||
More Information
Set Membership: |
|

G6PC2
Genomic Map