Assay Details
| Gene Symbol: HOXD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712302.2 | XP_002712348.1 | 1 - 2 | 597 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD12| Gene Symbol: HOXD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712302.2 | XP_002712348.1 | 1 - 2 | 597 | 89 | |
Set Membership: |
|