Assay Details
| Gene Symbol: LOC100351758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714849.2 | XP_002714895.1 | 1 - 2 | 129 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002714849.2 transcripts.
Set Membership: |
|

LOC100351758
Genomic Map