Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716748.2 | XP_002716794.1 | 8 - 9 | 1477 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRR3| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716748.2 | XP_002716794.1 | 8 - 9 | 1477 | 72 | |
Set Membership: |
|