Assay Details
| Gene Symbol: POU4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347269.1 | XP_017202758.1 | 1 | 22 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017347269.1 transcripts.
Set Membership: |
|

POU4F2
Genomic Map