Assay Details
| Gene Symbol: H1-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721618.3 | XP_002721664.1 | 314 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002721618.3 transcripts.
Set Membership: |
|

H1-5
Genomic Map