Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082168.2 | NP_001075637.2 | 1 | 610 | 109 | |
| GenBank mRNA | EB377152.1 | - | 2 | 392 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YWHAQ| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082168.2 | NP_001075637.2 | 1 | 610 | 109 | |
| GenBank mRNA | EB377152.1 | - | 2 | 392 | 109 | |
Set Membership: |
|