Assay Details
| Gene Symbol: SOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082627.2 | NP_001076096.2 | 3 - 4 | 365 | 92 | |
| XM_017347219.1 | XP_017202708.1 | 3 - 4 | 297 | 92 | ||
| GenBank mRNA | L12405.1 | - | 3 - 4 | 308 | 92 | |
| Z22644.1 | - | 308 | 92 | |||
More Information
Set Membership: |
|

SOD1
Genomic Map