Assay Details
| Gene Symbol: LOC4325457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766429.2 | XP_015621915.1 | 1 - 2 | 285 | 55 | |
| GenBank mRNA | AK072025.1 | - | 1 - 2 | 274 | 55 | |
| AK107219.1 | - | 1 - 2 | 258 | 55 | ||
| HQ013291.1 | - | 1 - 2 | 250 | 55 | ||
More Information
Set Membership: |
|

LOC4325457
Genomic Map