Assay Details
| Gene Symbol: LOC4325457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766429.2 | XP_015621915.1 | 6 - 7 | 850 | 74 | |
| XM_015766430.2 | XP_015621916.1 | 7 - 8 | 731 | 74 | ||
| GenBank mRNA | AK072025.1 | - | 6 - 7 | 839 | 74 | |
| AK107219.1 | - | 6 - 7 | 823 | 74 | ||
| HQ013291.1 | - | 6 - 7 | 815 | 74 | ||
More Information
Set Membership: |
|

LOC4325457
Genomic Map