Assay Details
| Gene Symbol: LOC4324061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765481.2 | XP_015620967.1 | 1 - 2 | 372 | 147 | |
| GenBank mRNA | AK073728.1 | - | 1 - 2 | 248 | 147 | |
| HQ324798.1 | - | 1 - 2 | 379 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765481.2 transcripts.
Set Membership: |
|

LOC4324061
Genomic Map