Assay Details
| Gene Symbol: LOC4324061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765481.2 | XP_015620967.1 | 5 - 6 | 1637 | 81 | |
| GenBank mRNA | AK073728.1 | - | 5 - 6 | 1513 | 81 | |
| HQ324798.1 | - | 5 - 6 | 1644 | 81 | ||
More Information
Set Membership: |
|

LOC4324061
Genomic Map