Assay Details
| Gene Symbol: LOC4324061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765481.2 | XP_015620967.1 | 6 - 7 | 1780 | 118 | |
| GenBank mRNA | AK073728.1 | - | 6 - 7 | 1656 | 118 | |
| HQ324798.1 | - | 6 - 7 | 1787 | 118 | ||
More Information
Set Membership: |
|

LOC4324061
Genomic Map