Assay Details
| Gene Symbol: LOC4327645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765933.2 | XP_015621419.1 | 1 - 2 | 197 | 112 | |
| XM_015765935.2 | XP_015621421.1 | 1 - 2 | 197 | 112 | ||
| XM_015765936.2 | XP_015621422.1 | 1 - 2 | 197 | 112 | ||
| GenBank mRNA | AK100326.1 | - | 1 - 2 | 197 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765933.2, XM_015765935.2, XM_015765936.2 transcripts.
Set Membership: |
|

LOC4327645
Genomic Map