Assay Details
| Gene Symbol: LOC4327645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765933.2 | XP_015621419.1 | 7 - 8 | 1212 | 150 | |
| XM_015765935.2 | XP_015621421.1 | 7 - 8 | 1059 | 150 | ||
| XM_015765936.2 | XP_015621422.1 | 7 - 8 | 1059 | 150 | ||
| GenBank mRNA | AK100326.1 | - | 7 - 8 | 1212 | 150 | |
| GQ267545.1 | - | 6 - 7 | 844 | 150 | ||
More Information
Set Membership: |
|

LOC4327645
Genomic Map