Assay Details
| Gene Symbol: LOC4327645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765933.2 | XP_015621419.1 | 8 - 9 | 1564 | 70 | |
| XM_015765935.2 | XP_015621421.1 | 8 - 9 | 1411 | 70 | ||
| XM_015765936.2 | XP_015621422.1 | 8 - 9 | 1411 | 70 | ||
| GenBank mRNA | AK100326.1 | - | 8 - 9 | 1564 | 70 | |
| GQ267545.1 | - | 7 - 8 | 1196 | 70 | ||
More Information
Set Membership: |
|

LOC4327645
Genomic Map