Assay Details
| Gene Symbol: LOC4325354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772328.2 | XP_015627814.1 | 4 - 5 | 469 | 66 | |
| GenBank mRNA | AK240913.1 | - | 4 - 5 | 476 | 66 | |
More Information
Set Membership: |
|

LOC4325354
Genomic Map