Assay Details
| Gene Symbol: LOC4325330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776740.2 | XP_015632226.1 | 3 - 4 | 741 | 59 | |
| GenBank mRNA | AY341853.1 | - | 3 - 4 | 647 | 59 | |
More Information
Set Membership: |
|

LOC4325330
Genomic Map