Assay Details
| Gene Symbol: LOC4325330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776740.2 | XP_015632226.1 | 7 - 8 | 1257 | 113 | |
| GenBank mRNA | AY341853.1 | - | 7 - 8 | 1163 | 113 | |
More Information
Set Membership: |
|

LOC4325330
Genomic Map