Assay Details
| Gene Symbol: LOC4326358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766465.1 | XP_015621951.1 | 3 - 4 | 1357 | 55 | |
| XM_015766466.1 | XP_015621952.1 | 4 - 5 | 1237 | 55 | ||
| GenBank mRNA | AK067076.1 | - | 3 - 4 | 1042 | 55 | |
| AK104302.1 | - | 3 - 4 | 986 | 55 | ||
More Information
Set Membership: |
|

LOC4326358
Genomic Map