Assay Details
| Gene Symbol: LOC4324370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765698.2 | XP_015621184.1 | 10 - 11 | 2641 | 87 | |
| XM_015765699.2 | XP_015621185.1 | 10 - 11 | 2636 | 87 | ||
| GenBank mRNA | AK121684.1 | - | 10 - 11 | 2642 | 87 | |
| BT132429.1 | - | 2 - 3 | 272 | 87 | ||
More Information
Set Membership: |
|

LOC4324370
Genomic Map