Assay Details
| Gene Symbol: LOC4324370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765698.2 | XP_015621184.1 | 12 - 13 | 2858 | 104 | |
| XM_015765699.2 | XP_015621185.1 | 12 - 13 | 2853 | 104 | ||
| GenBank mRNA | AK121684.1 | - | 12 - 13 | 2859 | 104 | |
| BT132429.1 | - | 4 - 5 | 489 | 104 | ||
More Information
Set Membership: |
|

LOC4324370
Genomic Map