Assay Details
| Gene Symbol: LOC4324370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765698.2 | XP_015621184.1 | 13 - 14 | 2959 | 89 | |
| XM_015765699.2 | XP_015621185.1 | 13 - 14 | 2954 | 89 | ||
| GenBank mRNA | AK121684.1 | - | 13 - 14 | 2960 | 89 | |
| BT132429.1 | - | 5 - 6 | 590 | 89 | ||
More Information
Set Membership: |
|

LOC4324370
Genomic Map