Assay Details
| Gene Symbol: LOC4324370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765698.2 | XP_015621184.1 | 1 - 2 | 674 | 105 | |
| XM_015765699.2 | XP_015621185.1 | 1 - 2 | 675 | 105 | ||
More Information
Set Membership: |
|

LOC4324370
Genomic Map