Assay Details
| Gene Symbol: LOC4324370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765698.2 | XP_015621184.1 | 8 - 9 | 2205 | 101 | |
| XM_015765699.2 | XP_015621185.1 | 8 - 9 | 2206 | 101 | ||
| GenBank mRNA | AK121684.1 | - | 8 - 9 | 2206 | 101 | |
More Information
Set Membership: |
|

LOC4324370
Genomic Map