Assay Details
| Gene Symbol: LOC4325168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795983.2 | XP_015651469.1 | 1 - 2 | 290 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325168| Gene Symbol: LOC4325168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795983.2 | XP_015651469.1 | 1 - 2 | 290 | 133 | |
Set Membership: |
|