Assay Details
| Gene Symbol: LOC4327289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756966.2 | XP_015612452.1 | 4 - 5 | 3900 | 92 | |
| GenBank mRNA | AK067157.1 | - | 4 - 5 | 1080 | 92 | |
More Information
Set Membership: |
|

LOC4327289
Genomic Map