Assay Details
| Gene Symbol: LOC4326988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766242.2 | XP_015621728.1 | 1 - 2 | 697 | 66 | |
| GenBank mRNA | AK070488.1 | - | 1 - 2 | 572 | 66 | |
More Information
Set Membership: |
|

LOC4326988
Genomic Map