Assay Details
| Gene Symbol: LOC4326988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766242.2 | XP_015621728.1 | 4 - 5 | 2700 | 73 | |
| GenBank mRNA | AK059365.1 | - | 3 - 4 | 692 | 73 | |
| AK070488.1 | - | 3 - 4 | 2956 | 73 | ||
More Information
Set Membership: |
|

LOC4326988
Genomic Map