Assay Details
| Gene Symbol: LOC4326316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765730.2 | XP_015621216.1 | 20 - 21 | 2342 | 114 | |
| GenBank mRNA | AK242763.1 | - | 20 - 21 | 2306 | 114 | |
More Information
Set Membership: |
|

LOC4326316
Genomic Map