Assay Details
| Gene Symbol: LOC4324190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765827.2 | XP_015621313.1 | 3 - 4 | 666 | 125 | |
| GenBank mRNA | AK064181.1 | - | 3 - 4 | 664 | 125 | |
| AK069131.1 | - | 3 - 4 | 614 | 125 | ||
| AK103451.1 | - | 3 - 4 | 575 | 125 | ||
More Information
Set Membership: |
|

LOC4324190
Genomic Map