Assay Details
| Gene Symbol: LOC4327728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792836.2 | XP_015648322.1 | 16 - 17 | 3519 | 91 | |
| GenBank mRNA | AK058981.1 | - | 2 - 3 | 284 | 91 | |
| AK100135.1 | - | 15 - 16 | 2838 | 91 | ||
More Information
Set Membership: |
|

LOC4327728
Genomic Map