Assay Details
| Gene Symbol: LOC4327728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792836.2 | XP_015648322.1 | 19 - 20 | 4050 | 62 | |
| GenBank mRNA | AK058981.1 | - | 5 - 6 | 815 | 62 | |
| AK100135.1 | - | 18 - 19 | 3370 | 62 | ||
More Information
Set Membership: |
|

LOC4327728
Genomic Map