Assay Details
| Gene Symbol: LOC4325312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767352.2 | XP_015622838.1 | 1 - 2 | 161 | 73 | |
| GenBank mRNA | AB015288.1 | - | 1 - 2 | 72 | 73 | |
| AB015971.1 | - | 1 - 2 | 76 | 73 | ||
| AK072731.1 | - | 1 - 2 | 95 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767352.2 transcripts.
Set Membership: |
|

LOC4325312
Genomic Map