Assay Details
| Gene Symbol: LOC4325312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767352.2 | XP_015622838.1 | 4 - 5 | 395 | 90 | |
| GenBank mRNA | AB015288.1 | - | 4 - 5 | 306 | 90 | |
| AB015971.1 | - | 4 - 5 | 310 | 90 | ||
More Information
Set Membership: |
|

LOC4325312
Genomic Map