Assay Details
| Gene Symbol: LOC4325312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767352.2 | XP_015622838.1 | 7 - 8 | 751 | 65 | |
| GenBank mRNA | AB015288.1 | - | 7 - 8 | 662 | 65 | |
| AB015971.1 | - | 7 - 8 | 666 | 65 | ||
| AK072731.1 | - | 7 - 8 | 685 | 65 | ||
More Information
Set Membership: |
|

LOC4325312
Genomic Map