Assay Details
| Gene Symbol: LOC4326608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776785.2 | XP_015632271.1 | 14 - 15 | 2142 | 63 | |
| XM_015776864.2 | XP_015632350.1 | 12 - 13 | 2052 | 63 | ||
| GenBank mRNA | AK073890.1 | - | 14 - 15 | 2148 | 63 | |
| AY693783.1 | - | 9 - 10 | 1054 | 63 | ||
More Information
Set Membership: |
|

LOC4326608
Genomic Map