Assay Details
| Gene Symbol: LOC4326684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765426.2 | XP_015620912.1 | 9 - 10 | 1506 | 81 | |
| GenBank mRNA | AK060383.1 | - | 1 - 2 | 316 | 81 | |
More Information
Set Membership: |
|

LOC4326684
Genomic Map