Assay Details
Gene Symbol: LOC4327860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766422.2 | XP_015621908.1 | 2 - 3 | 837 | 110 | |
GenBank mRNA | AK071667.1 | - | 2 - 3 | 650 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
