Assay Details
| Gene Symbol: LOC4325901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766649.2 | XP_015622135.1 | 10 - 11 | 1344 | 76 | |
| GenBank mRNA | AB257288.1 | - | 10 - 11 | 1112 | 76 | |
| AK121375.1 | - | 10 - 11 | 1248 | 76 | ||
More Information
Set Membership: |
|

LOC4325901
Genomic Map